accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO(R)(R)H3N

CAS RN 28250-14-8

IUPAC Name: (1R,2R)-2-(ethoxycarbonyl)cyclohexan-1-aminium
Synonyms: (1R,2R)-2-(ethoxycarbonyl)cyclohexan-1-aminium
Molecular Weight (g/mol): 172.25
Molecular Formula: C9H18NO2
InChi Key: VODUKXHGDCJEOZ-HTQZYQBOSA-O
SMILES: CCOC(=O)[C@@H]1CCCC[C@H]1[NH3+]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)