accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNONNH2HCl

CAS RN 2827-56-7

IUPAC Name: 1-aminoimidazolidine-2,4-dione hydrochloride
Synonyms: 1-aminoimidazolidine-2,4-dione hydrochloride
Molecular Weight (g/mol): 151.55
Molecular Formula: C3H6ClN3O2
InChi Key: WEOHANUVLKERQI-UHFFFAOYSA-N
SMILES: Cl.NN1CC(=O)NC1=O
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)
Physical Form 
  • (4)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)