accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2829-42-7

IUPAC Name: disodium (3E)-3-[2-(4-{[(4-{2-[(1E)-3-carboxylato-4-oxocyclohexa-2,5-dien-1-ylidene]hydrazin-1-yl}phenyl)carbamoyl]amino}phenyl)hydrazin-1-ylidene]-6-oxocyclohexa-1,4-diene-1-carboxylate
Synonyms: disodium (3E)-3-[2-(4-{[(4-{2-[(1E)-3-carboxylato-4-oxocyclohexa-2,5-dien-1-ylidene]hydrazin-1-yl}phenyl)carbamoyl]amino}phenyl)hydrazin-1-ylidene]-6-oxocyclohexa-1,4-diene-1-carboxylate
Molecular Weight (g/mol): 584.46
Molecular Formula: C27H18N6Na2O7
InChi Key: NPQLRSWJRSHFMF-CTLWQRGVSA-L
SMILES: [Na+].[Na+].[O-]C(=O)C1=C\C(C=CC1=O)=N\NC1=CC=C(NC(=O)NC2=CC=C(N\N=C3/C=CC(=O)C(=C3)C([O-])=O)C=C2)C=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results