accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOSOOONHHNHNOHNNHHNONHSOHOOSOHOOSOHOONH

CAS RN 2829-43-8

IUPAC Name: 6-imino-5-(2-{4-[({4-[2-(2-imino-8-oxo-6-sulfo-2,8-dihydronaphthalen-1-yl)hydrazin-1-yl]-3-sulfophenyl}carbamoyl)amino]-2-sulfophenyl}hydrazin-1-yl)-4-oxo-4,6-dihydronaphthalene-2-sulfonic acid
Synonyms: 6-imino-5-(2-{4-[({4-[2-(2-imino-8-oxo-6-sulfonaphthalen-1-yl)hydrazin-1-yl]-3-sulfophenyl}carbamoyl)amino]-2-sulfophenyl}hydrazin-1-yl)-4-oxonaphthalene-2-sulfonic acid
Molecular Weight (g/mol): 902.85
Molecular Formula: C33H26N8O15S4
InChi Key: ZSJSLMQZBXTJJO-UHFFFAOYSA-N
SMILES: OS(=O)(=O)C1=CC(=O)C2=C(NNC3=CC=C(NC(=O)NC4=CC=C(NNC5=C6C(=O)C=C(C=C6C=CC5=N)S(O)(=O)=O)C(=C4)S(O)(=O)=O)C=C3S(O)(=O)=O)C(=N)C=CC2=C1
Color 
  • (2)
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)