accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOBOOCH2

CAS RN 283-56-7

IUPAC Name: 2,8,9-trioxa-5-aza-1-borabicyclo[3.3.3]undecane
Synonyms: 2,8,9-trioxa-5-aza-1-borabicyclo[3.3.3]undecane
Molecular Weight (g/mol): 156.98
Molecular Formula: C6H12BNO3
InChi Key: NKPKVNRBHXOADG-UHFFFAOYSA-N
SMILES: C1CN2CCOB(O1)OCC2
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Grade 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)