accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 28302-36-5

IUPAC Name: copper(2+) trisodium (14S,15S)-14-(2-carboxylatoethyl)-12-(carboxylatomethyl)-20-ethenyl-5-ethyl-4,9,15,19-tetramethyl-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1(20),2,4,6(24),7,9,11(23),12,16,18-decaene-21,22-diide-10-carboxylate
Synonyms: copper(2+) trisodium (14S,15S)-14-(2-carboxylatoethyl)-12-(carboxylatomethyl)-20-ethenyl-5-ethyl-4,9,15,19-tetramethyl-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1(20),2,4,6(24),7,9,11(23),12,16,18-decaene-21,22-diide-10-carboxylate
Molecular Weight (g/mol): 724.16
Molecular Formula: C34H31CuN4Na3O6
InChi Key: HWDGVJUIHRPKFR-ZWPRWVNUSA-I
SMILES: [Na+].[Na+].[Na+].[Cu++].CCC1=C(C)C2=CC3=C(C=C)C(C)=C([N-]3)C=C3[N-]C([C@@H](CCC([O-])=O)[C@@H]3C)=C(CC([O-])=O)C3=NC(=CC1=N2)C(C)=C3C([O-])=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results