accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOFFF

CAS RN 28314-80-9

IUPAC Name: 2,4,6-trifluorobenzoate
Synonyms: 2,4,6-trifluorobenzoate
Molecular Weight (g/mol): 175.09
Molecular Formula: C7H2F3O2
InChi Key: SJZATRRXUILGHH-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=C(F)C=C(F)C=C1F
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)