accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CBrCH3

CAS RN 28320-31-2

IUPAC Name: 2-bromo-9,9-dimethyl-9H-fluorene
Synonyms: 2-bromo-9,9-dimethylfluorene
Molecular Weight (g/mol): 273.17
Molecular Formula: C15H13Br
InChi Key: MBHPOBSZPYEADG-UHFFFAOYSA-N
SMILES: CC1(C)C2=CC=CC=C2C2=C1C=C(Br)C=C2
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)