accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOBr

CAS RN 2834-05-1

IUPAC Name: 11-bromoundecanoic acid
Synonyms: undecanoic acid, 11-bromo-
Molecular Weight (g/mol): 265.19
Molecular Formula: C11H21BrO2
InChi Key: IUDGNRWYNOEIKF-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCCCCCCBr
Percent Purity 
  • (3)
  • (2)
  • (3)
Physical Form 
  • (3)
  • (2)
  • (3)
Quantity 
  • (1)
  • (3)
  • (3)
  • (1)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (3)