accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2O

CAS RN 2835-77-0

IUPAC Name: 2-benzoylaniline
Synonyms: 2-benzoylaniline
Molecular Weight (g/mol): 197.24
Molecular Formula: C13H11NO
InChi Key: MAOBFOXLCJIFLV-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1C(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (7)
Percent Purity 
  • (2)
  • (7)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (7)
  • (2)
  • (7)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)