accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaHOOO

CAS RN 2836-32-0

IUPAC Name: sodium 2-hydroxyacetate
Synonyms: sodium glycolate
Molecular Weight (g/mol): 98.03
Molecular Formula: C2H3NaO3
InChi Key: VILMUCRZVVVJCA-UHFFFAOYSA-M
SMILES: [Na+].OCC([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (3)
  • (5)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)