accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 28418-86-2

IUPAC Name: 1-(4-methylnaphthalen-1-yl)ethan-1-one
Synonyms: 1-(4-methylnaphthalen-1-yl)ethanone
Molecular Weight (g/mol): 184.24
Molecular Formula: C13H12O
InChi Key: BKGIZYOJHJKFJP-UHFFFAOYSA-N
SMILES: CC(=O)C1=C2C=CC=CC2=C(C)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results