accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2845-78-5

IUPAC Name: 4-bromo-5-phenyl-1H-pyrazol-3-amine
Synonyms: 4-bromo-5-phenyl-1H-pyrazol-3-amine
Molecular Weight (g/mol): 238.09
Molecular Formula: C9H8BrN3
InChi Key: QTNVXMOPTHGCII-UHFFFAOYSA-N
SMILES: NC1=NNC(=C1Br)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results