accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 28460-01-7

IUPAC Name: diethyl [(methylsulfanyl)methyl]phosphonate
Synonyms: diethyl (methylsulfanyl)methylphosphonate
Molecular Weight (g/mol): 198.22
Molecular Formula: C6H15O3PS
InChi Key: RAABCCZMKIIGJW-UHFFFAOYSA-N
SMILES: CCOP(=O)(CSC)OCC
Boiling Point 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results