accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 28469-92-3

IUPAC Name: 1,3-dichloro-2-ethenylbenzene
Synonyms: 2,6-dichlorostyrene
Molecular Weight (g/mol): 173.04
Molecular Formula: C8H6Cl2
InChi Key: YJCVRMIJBXTMNR-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(Cl)=C1C=C
Boiling Point 
  • (3)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)