CAS RN 28503-70-0
IUPAC Name:
titanium(4+) tetrakis(2-methylbenzen-1-olate)
Synonyms:
titanium(4+) tetrakis(2-methylbenzenolate)
Molecular Weight (g/mol):
476.4
Molecular Formula:
C28H28O4Ti
InChi Key:
RLMZHOIDXMRYMJ-UHFFFAOYSA-J
SMILES:
[Ti+4].CC1=CC=CC=C1[O-].CC1=CC=CC=C1[O-].CC1=CC=CC=C1[O-].CC1=CC=CC=C1[O-]
1
–
1
of
1
results
1
Titanium(IV) methylphenoxide, tech., Thermo Scientific™
CAS: 28503-70-0 Molecular Formula: C28H28O4Ti MDL Number: MFCD00061534 Synonym: Titanium(IV) cresylate