accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COPSOCH3ONClClCl

CAS RN 285138-81-0

IUPAC Name: O,O-diethyl O-3,5,6-trichloropyridin-2-yl phosphorothioate
Synonyms: terial suSCon Green piridane killmaster equity radar (fungicide) m-chlorpyrifos zidil pyrinex grofo
Molecular Weight (g/mol): 350.57
Molecular Formula: C9H11Cl3NO3PS
InChi Key: SBPBAQFWLVIOKP-UHFFFAOYSA-N
SMILES: CCOP(=S)(OCC)OC1=NC(Cl)=C(Cl)C=C1Cl
Grade 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)