accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 28519-50-8

IUPAC Name: potassium [(4-methylphenyl)oxo-λ⁶-disulfanyl]olate
Synonyms: potassium (4-methylphenyl)oxo-λ⁶-disulfanylolate
Molecular Weight (g/mol): 226.35
Molecular Formula: C7H7KO2S2
InChi Key: RUDNWZFWWJFUSF-UHFFFAOYSA-M
SMILES: [K+].CC1=CC=C(C=C1)S([O-])(=O)=S
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)