accessibility menu, dialog, popup

UserName

Viewing profile as user:
OONNOO

CAS RN 28537-70-4

IUPAC Name: 1-[4-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)butyl]-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 1-[4-(2,5-dioxopyrrol-1-yl)butyl]pyrrole-2,5-dione
Molecular Weight (g/mol): 248.24
Molecular Formula: C12H12N2O4
InChi Key: WXXSHAKLDCERGU-UHFFFAOYSA-N
SMILES: O=C1C=CC(=O)N1CCCCN1C(=O)C=CC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results