accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOOOHOHOHHO

CAS RN 28541-83-5

IUPAC Name: 4-methyl-7-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2H-chromen-2-one
Synonyms: 4methylumbelliferylbdglucopyranoside
Molecular Weight (g/mol): 338.31
Molecular Formula: C16H18O8
InChi Key: YUDPTGPSBJVHCN-UHFFFAOYNA-N
SMILES: CC1=CC(=O)OC2=CC(OC3OC(CO)C(O)C(O)C3O)=CC=C12
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)