accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2855-13-2

IUPAC Name: 3-(aminomethyl)-3,5,5-trimethylcyclohexan-1-amine
Synonyms: isophorone diamine
Molecular Weight (g/mol): 170.3
Molecular Formula: C10H22N2
InChi Key: RNLHGQLZWXBQNY-UHFFFAOYNA-N
SMILES: CC1(C)CC(N)CC(C)(CN)C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (5)
  • (1)
  • (4)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)