accessibility menu, dialog, popup

UserName

Viewing profile as user:
O(S)HNOO

CAS RN 28562-58-5

IUPAC Name: (2S)-4-oxoazetidin-2-yl benzoate
Synonyms: (2S)-4-oxoazetidin-2-yl benzoate
Molecular Weight (g/mol): 191.19
Molecular Formula: C10H9NO3
InChi Key: HJJGOOONOIFDRH-VIFPVBQESA-N
SMILES: O=C(O[C@H]1CC(=O)N1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results