accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 28562-68-7

IUPAC Name: 3-methyloxetane-3-carboxylic acid
Synonyms: 3-methyloxetane-3-carboxylic acid
Molecular Weight (g/mol): 116.12
Molecular Formula: C5H8O3
InChi Key: PMPZEEUNGQSAIQ-UHFFFAOYSA-N
SMILES: CC1(COC1)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)