CAS RN 28598-81-4
IUPAC Name:
ethyl 9-bromononanoate
Synonyms:
ethyl 9-bromononanoate
Molecular Weight (g/mol):
265.19
Molecular Formula:
C11H21BrO2
InChi Key:
KXZBPLOPBKIUTC-UHFFFAOYSA-N
SMILES:
CCOC(=O)CCCCCCCCBr
1
–
1
of
1
results
1
Ethyl 9-Bromononanoate 95.0+%, TCI America™
CAS: 28598-81-4 Molecular Formula: C11H21BrO2 Molecular Weight (g/mol): 265.191 MDL Number: MFCD06797065 InChI Key: KXZBPLOPBKIUTC-UHFFFAOYSA-N Synonym: 9-Bromononanoic Acid Ethyl Ester PubChem CID: 560317 IUPAC Name: ethyl 9-bromononanoate SMILES: CCOC(=O)CCCCCCCCBr