accessibility menu, dialog, popup

UserName

Viewing profile as user:
O

CAS RN 286-99-7

IUPAC Name: 13-oxabicyclo[10.1.0]tridecane
Synonyms: cyclododecane epoxide epoxycyclododecane cyclododeceneepoxide
Molecular Weight (g/mol): 182.31
Molecular Formula: C12H22O
InChi Key: VLJLXEKIAALSJE-UHFFFAOYNA-N
SMILES: C1CCCCCC2OC2CCCC1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)