accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 286014-42-4

IUPAC Name: 1,3-bis(adamantan-1-yl)-3H-1λ⁵-imidazol-1-ylium; tetrafluoroboranuide
Synonyms: 1,3-bis(adamantan-1-yl)-1λ⁵-imidazol-1-ylium tetrafluoroborate
Molecular Weight (g/mol): 424.33
Molecular Formula: C23H33BF4N2
InChi Key: KVWCCJYLKCSVME-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.C1C2CC3CC1CC(C2)(C3)N1C=C[N+](=C1)C12CC3CC(CC(C3)C1)C2
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results