accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOOOHOONH2

CAS RN 28607-46-7

IUPAC Name: 4-methylbenzene-1-sulfonic acid; benzyl 2-amino-3-phenylpropanoate
Synonyms: benzyl 2-amino-3-phenylpropanoate; para-toluene sulfonate
Molecular Weight (g/mol): 427.52
Molecular Formula: C23H25NO5S
InChi Key: ZLZGBBIPWXUQST-UHFFFAOYNA-N
SMILES: CC1=CC=C(C=C1)S(O)(=O)=O.NC(CC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)