accessibility menu, dialog, popup

UserName

Viewing profile as user:
OFFFBrF

CAS RN 286932-57-8

IUPAC Name: 2-bromo-1-fluoro-4-(trifluoromethoxy)benzene
Synonyms: 2-bromo-1-fluoro-4-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 259
Molecular Formula: C7H3BrF4O
InChi Key: VVVFPEJORKPEST-UHFFFAOYSA-N
SMILES: FC1=C(Br)C=C(OC(F)(F)F)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results