accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOCH3OOCH3O

CAS RN 2873-29-2

IUPAC Name: [3,4-bis(acetyloxy)-3,4-dihydro-2H-pyran-2-yl]methyl acetate
Synonyms: [3,4-bis(acetyloxy)-3,4-dihydro-2H-pyran-2-yl]methyl acetate
Molecular Weight (g/mol): 272.25
Molecular Formula: C12H16O7
InChi Key: LLPWGHLVUPBSLP-UHFFFAOYNA-N
SMILES: CC(=O)OCC1OC=CC(OC(C)=O)C1OC(C)=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)