accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 28733-43-9

IUPAC Name: 5-bromopyridine-3-carboxamide
Synonyms: 5-bromonicotinamide C6H5BrN2O
Molecular Weight (g/mol): 201.02
Molecular Formula: C6H5BrN2O
InChi Key: YOQRXZIMSKLRCY-UHFFFAOYSA-N
SMILES: NC(=O)C1=CN=CC(Br)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results