accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 28752-82-1

IUPAC Name: 2-(prop-2-en-1-yloxy)benzaldehyde
Synonyms: 2-(prop-2-en-1-yloxy)benzaldehyde
Molecular Weight (g/mol): 162.19
Molecular Formula: C10H10O2
InChi Key: BXCJDECTRRMSCV-UHFFFAOYSA-N
SMILES: C=CCOC1=CC=CC=C1C=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results