accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONONOO

CAS RN 28768-32-3

IUPAC Name: 4-[(4-{bis[(oxiran-2-yl)methyl]amino}phenyl)methyl]-N,N-bis[(oxiran-2-yl)methyl]aniline
Synonyms: 4-({4-[bis(oxiran-2-ylmethyl)amino]phenyl}methyl)-N,N-bis(oxiran-2-ylmethyl)aniline
Molecular Weight (g/mol): 422.53
Molecular Formula: C25H30N2O4
InChi Key: FAUAZXVRLVIARB-UHFFFAOYNA-N
SMILES: C(C1CO1)N(CC1CO1)C1=CC=C(CC2=CC=C(C=C2)N(CC2CO2)CC2CO2)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)