accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 28788-62-7

IUPAC Name: 4-butylbenzoyl chloride
Synonyms: 4-butylbenzoyl chloride
Molecular Weight (g/mol): 196.67
Molecular Formula: C11H13ClO
InChi Key: OUOWCSJYDCPVDM-UHFFFAOYSA-N
SMILES: CCCCC1=CC=C(C=C1)C(Cl)=O
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)