accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 28791-97-1

CH3NNNNH3CCH3NNCH3H3CCH3

CAS RN 28791-97-1

IUPAC Name: 1-[bis(3,5-dimethyl-1H-pyrazol-1-yl)methyl]-3,5-dimethyl-1H-pyrazole
Synonyms: 1-[bis(3,5-dimethylpyrazol-1-yl)methyl]-3,5-dimethylpyrazole
Molecular Weight (g/mol): 298.39
Molecular Formula: C16H22N6
InChi Key: COKUMKYWINNWLV-UHFFFAOYSA-N
SMILES: CC1=NN(C(N2N=C(C)C=C2C)N2N=C(C)C=C2C)C(C)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results