accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CO

CAS RN 287944-16-5

IUPAC Name: 2-(3,6-dihydro-2H-pyran-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(3,6-dihydro-2H-pyran-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 210.08
Molecular Formula: C11H19BO3
InChi Key: DOSGEBYQRMBTGS-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CCOCC1
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)