accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 288071-87-4

IUPAC Name: 4,7-bis(5-bromothiophen-2-yl)-2,1,3-benzothiadiazole
Synonyms: 4,7-bis(5-bromothiophen-2-yl)-2,1,3-benzothiadiazole
Molecular Weight (g/mol): 458.2
Molecular Formula: C14H6Br2N2S3
InChi Key: ZIIMIGRZSUYQGW-UHFFFAOYSA-N
SMILES: BrC1=CC=C(S1)C1=CC=C(C2=CC=C(Br)S2)C2=NSN=C12
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)