accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 288148-34-5

CH3NNSOOCl

CAS RN 288148-34-5

IUPAC Name: 1-methyl-1H-pyrazole-4-sulfonyl chloride
Synonyms: 1-methylpyrazole-4-sulfonyl chloride
Molecular Weight (g/mol): 180.61
Molecular Formula: C4H5ClN2O2S
InChi Key: RDAVKKQKMLINOH-UHFFFAOYSA-N
SMILES: CN1C=C(C=N1)S(Cl)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results