accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOBr

CAS RN 2882-19-1

IUPAC Name: ethyl 2-bromo-2-phenylacetate
Synonyms: ethyl 2-bromo-2-phenylacetate
Molecular Weight (g/mol): 243.1
Molecular Formula: C10H11BrO2
InChi Key: BKTKLDMYHTUESO-UHFFFAOYNA-N
SMILES: CCOC(=O)C(Br)C1=CC=CC=C1
Boiling Point 
  • (3)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (6)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (2)
  • (3)
  • (2)
  • (6)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)