accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 288252-38-0

IUPAC Name: 1-(4-chlorophenyl)-5-methyl-1H-pyrazole-4-carbonyl chloride
Synonyms: 1-(4-chlorophenyl)-5-methylpyrazole-4-carbonyl chloride
Molecular Weight (g/mol): 255.1
Molecular Formula: C11H8Cl2N2O
InChi Key: KUQHQFYKJPXTNB-UHFFFAOYSA-N
SMILES: CC1=C(C=NN1C1=CC=C(Cl)C=C1)C(Cl)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results