accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C

CAS RN 2883-02-5

IUPAC Name: nonylcyclohexane
Synonyms: N-nonylcyclohexane
Molecular Weight (g/mol): 210.41
Molecular Formula: C15H30
InChi Key: CLMFECCMAVQYQA-UHFFFAOYSA-N
SMILES: CCCCCCCCCC1CCCCC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results