accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3CH3OCH3(Z)

CAS RN 2883-98-9

IUPAC Name: 1,2,4-trimethoxy-5-[(1Z)-prop-1-en-1-yl]benzene
Synonyms: β-asarone cis-asarone cis-isoasarone trans-asarone cis-isoelemicin (Z)-asarone
Molecular Weight (g/mol): 208.26
Molecular Formula: C12H16O3
InChi Key: RKFAZBXYICVSKP-WAYWQWQTSA-N
SMILES: COC1=CC(OC)=C(\C=C/C)C=C1OC
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)