accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 288315-03-7

IUPAC Name: 3,3-difluoroazetidine hydrochloride
Synonyms: 3,3-difluoroazetidine hydrochloride
Molecular Weight (g/mol): 129.53
Molecular Formula: C3H6ClF2N
InChi Key: CDBAEFXTCRKJPZ-UHFFFAOYSA-N
SMILES: Cl.FC1(F)CNC1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)