CAS RN 288617-78-7
CAS RN 288617-78-7
IUPAC Name:
2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-2-methyloct-7-enoic acid
Synonyms:
2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-2-methyloct-7-enoic acid
Molecular Weight (g/mol):
393.48
Molecular Formula:
C24H27NO4
InChi Key:
QVNDFQWCJFAEFY-XMMPIXPASA-N
SMILES:
CC(CCCCC=C)(NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
1
–
1
of
1
results
1
Advanced Chem Tech Fmoc- R -2- 5-Hexenyl alanine, Advanced ChemTech
CAS: 288617-78-7 Molecular Formula: C24H27NO4 Molecular Weight (g/mol): 393.48 MDL Number: MFCD28009658 InChI Key: QVNDFQWCJFAEFY-XMMPIXPASA-N IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-2-methyloct-7-enoic acid SMILES: CC(CCCCC=C)(NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O