accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOClOOCH3

CAS RN 28868-76-0

IUPAC Name: 1,3-dimethyl 2-chloropropanedioate
Synonyms: 1,3-dimethyl 2-chloropropanedioate
Molecular Weight (g/mol): 166.56
Molecular Formula: C5H7ClO4
InChi Key: LNBQBURECUEBKZ-UHFFFAOYSA-N
SMILES: COC(=O)C(Cl)C(=O)OC
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results