CAS RN 289039-85-6
IUPAC Name:
methyl 3-chloro-5-iodobenzoate
Synonyms:
methyl 3-chloro-5-iodobenzoate
Molecular Weight (g/mol):
296.49
Molecular Formula:
C8H6ClIO2
InChi Key:
ZDSIXLJPASGHDW-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC(Cl)=CC(I)=C1
1
–
1
of
1
results
1
Promotions Available
Methyl 3-chloro-5-iodobenzoate, 98%, Thermo Scientific Chemicals
Methyl 3-chloro-5-iodobenzoate, 98%, Thermo Scientific Chemicals
CAS: 289039-85-6 Molecular Formula: C8H6ClIO2 Molecular Weight (g/mol): 296.488 MDL Number: MFCD00673018 InChI Key: ZDSIXLJPASGHDW-UHFFFAOYSA-N Synonym: acmc-20akhf,methyl 5-chloro-3-iodobenzoate,3-chloro-5-iodo-benzoic acid methyl ester,3-chloro-5-iodobenzoic acid methyl ester PubChem CID: 24721134 IUPAC Name: methyl 3-chloro-5-iodobenzoate SMILES: COC(=O)C1=CC(=CC(=C1)I)Cl