accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2892-29-7

IUPAC Name: 1-(3-chloro-4-hydroxyphenyl)ethan-1-one
Synonyms: 1-(3-chloro-4-hydroxyphenyl)ethanone
Molecular Weight (g/mol): 170.59
Molecular Formula: C8H7ClO2
InChi Key: GMTSPBYBJKGPJF-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(O)C(Cl)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results