accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 28921-35-9

IUPAC Name: ethyl 2-methylfuran-3-carboxylate
Synonyms: ethyl 2-methylfuran-3-carboxylate
Molecular Weight (g/mol): 154.17
Molecular Formula: C8H10O3
InChi Key: DOHUEYINVDLUSG-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)OC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results