accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 28924-21-2

IUPAC Name: 1-[3,5-bis(benzyloxy)phenyl]ethan-1-one
Synonyms: 1-[3,5-bis(benzyloxy)phenyl]ethanone
Molecular Weight (g/mol): 332.4
Molecular Formula: C22H20O3
InChi Key: KOJXGMJOTRYLBD-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(OCC2=CC=CC=C2)=CC(OCC2=CC=CC=C2)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results