accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2OCl

CAS RN 2894-51-1

IUPAC Name: 2-(4-chlorobenzoyl)aniline
Synonyms: 2-(4-chlorobenzoyl)aniline
Molecular Weight (g/mol): 231.68
Molecular Formula: C13H10ClNO
InChi Key: APHLSUBLNQBFTM-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1C(=O)C1=CC=C(Cl)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results