CAS RN 28957-04-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Oridonin, R. rubescens
A cell-permeable diterpenoid compound that possesses anti NF-κB activity and displays antiproliferative and antiangiogenic properties.